siRNA Details
| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153311.1 | NP_695223.1 | 5 | 515 | ||
| XM_008766194.1 | XP_008764416.1 | 3 | 378 | |||
| GenBank mRNA | AF537098.1 | - | 5 | 515 | ||
| AF537099.1 | - | 3 | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss5| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153311.1 | NP_695223.1 | 5 | 515 | ||
| XM_008766194.1 | XP_008764416.1 | 3 | 378 | |||
| GenBank mRNA | AF537098.1 | - | 5 | 515 | ||
| AF537099.1 | - | 3 | 293 | |||