siRNA Details
| Gene Symbol: Cyp4f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153318.1 | NP_695230.1 | 2 | 209 | ||
| XM_006241049.1 | XP_006241111.1 | 2 | 283 | |||
| XM_008765156.1 | XP_008763378.1 | 2 | 362 | |||
| GenBank mRNA | BC078713.1 | - | 2 | 119 | 7107801 | |
| U39208.1 | - | 2 | 209 | |||

Cyp4f6
Genomic Map