siRNA Details
| Gene Symbol: Ppp1r17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153467.1 | NP_703197.1 | 5 | 560 | ||
| XM_006236545.2 | XP_006236607.1 | 5 | 1069 | |||
| XM_008762878.1 | XP_008761100.1 | 6 | 996 | |||
| GenBank mRNA | AF294688.1 | - | 5 | 560 | ||
| FQ213688.1 | - | 5 | 522 | |||

Ppp1r17
Genomic Map