siRNA Details
| Gene Symbol: Ppp1r17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153467.1 | NP_703197.1 | 3 | 335 | ||
| XM_006236545.2 | XP_006236607.1 | 3 | 844 | |||
| XM_008762878.1 | XP_008761100.1 | 4 | 771 | |||
| GenBank mRNA | AF294688.1 | - | 3 | 335 | ||
| FQ213688.1 | - | 3 | 297 | |||

Ppp1r17
Genomic Map