siRNA Details
| Gene Symbol: Lzts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153470.1 | NP_703200.1 | 4 | 2005 | ||
| GenBank mRNA | AB075607.1 | - | 4 | 2005 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lzts1| Gene Symbol: Lzts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153470.1 | NP_703200.1 | 4 | 2005 | ||
| GenBank mRNA | AB075607.1 | - | 4 | 2005 | ||