siRNA Details
Gene Symbol: Rnf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153471.1 | NP_703201.1 | 16 | 2483 | ||
XM_006230219.2 | XP_006230281.1 | 16 | 2561 | |||
GenBank mRNA | AF352815.1 | - | 16 | 2483 |
