siRNA Details
Gene Symbol: Rnf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153471.1 | NP_703201.1 | 9 | 1149 | ||
GenBank mRNA | AF352815.1 | - | 9 | 1149 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153471.1 | NP_703201.1 | 9 | 1149 | ||
GenBank mRNA | AF352815.1 | - | 9 | 1149 |