siRNA Details
Gene Symbol: Myo7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153473.1 | NP_703203.1 | 47 | 6534 | ||
XM_006229742.2 | XP_006229804.1 | 46 | 7454 | |||
XM_006229743.2 | XP_006229805.1 | 46 | 7339 | |||
XM_006229744.2 | XP_006229806.1 | 47 | 6682 | |||
GenBank mRNA | AB091825.1 | - | 47 | 6534 |
