siRNA Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270871.1 | NP_001257800.1 | 14 | 1162 | ||
| NM_001270872.1 | NP_001257801.1 | 13 | 1126 | |||
| NM_181366.2 | NP_852031.1 | 15 | 1210 | |||
| XM_006256916.2 | XP_006256978.1 | 15 | 1250 | |||
| XM_008773154.1 | XP_008771376.1 | 14 | 1590 | |||
| XM_008773155.1 | XP_008771377.1 | 14 | 1200 | |||
| XM_008773156.1 | XP_008771378.1 | 13 | 1554 | |||
| XM_008773157.1 | XP_008771379.1 | 13 | 1548 | |||
| XM_008773158.1 | XP_008771380.1 | 12 | 1512 | |||
| XM_008773159.1 | XP_008771381.1 | 11 | 1103 | |||
| XM_008773160.1 | XP_008771382.1 | 14 | 1591 | |||
| GenBank mRNA | AF538953.1 | - | 15 | 1220 | ||
| AF538958.1 | - | 14 | 1172 | |||
| AF538959.1 | - | 13 | 1136 | |||
More Information
Set Membership: |
|

Gpr64
Genomic Map