siRNA Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153628.2 | NP_705892.1 | 6 | 607 | ||
| GenBank mRNA | AB022714.2 | - | 6 | 608 | ||
| FQ214257.1 | - | 3 | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec11c| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153628.2 | NP_705892.1 | 6 | 607 | ||
| GenBank mRNA | AB022714.2 | - | 6 | 608 | ||
| FQ214257.1 | - | 3 | 127 | |||