siRNA Details
| Gene Symbol: Hspa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153629.1 | NP_705893.1 | 4 | 615 | ||
| XM_006246232.1 | XP_006246294.1 | 4 | 659 | |||
| GenBank mRNA | AF077354.1 | - | 4 | 615 | ||
| FQ222433.1 | - | 5 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa4| Gene Symbol: Hspa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153629.1 | NP_705893.1 | 4 | 615 | ||
| XM_006246232.1 | XP_006246294.1 | 4 | 659 | |||
| GenBank mRNA | AF077354.1 | - | 4 | 615 | ||
| FQ222433.1 | - | 5 | 823 | |||