siRNA Details
| Gene Symbol: Eaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172047.1 | NP_742044.1 | 5 | 697 | ||
| XM_008768707.1 | XP_008766929.1 | 5 | 584 | |||
| XM_008768708.1 | XP_008766930.1 | 3 | 1253 | |||
| XM_008768709.1 | XP_008766931.1 | 3 | 983 | |||
| GenBank mRNA | AY049022.1 | - | 5 | 697 | ||

Eaf2
Genomic Map