siRNA Details
| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007606.1 | NP_001007607.1 | 7, 8 | 1016 | ||
| XM_006233136.2 | XP_006233198.1 | 7, 8 | 1051 | |||
| GenBank mRNA | AY145052.1 | - | 3, 4 | 443 | ||
| BC061765.1 | - | 7, 8 | 1016 | 5620893 | ||

Sars
Genomic Map