siRNA Details
| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007606.1 | NP_001007607.1 | 1, 2 | 199 | ||
| XM_006233136.2 | XP_006233198.1 | 1, 2 | 234 | |||
| GenBank mRNA | BC061765.1 | - | 1, 2 | 199 | 5620893 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sars| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007606.1 | NP_001007607.1 | 1, 2 | 199 | ||
| XM_006233136.2 | XP_006233198.1 | 1, 2 | 234 | |||
| GenBank mRNA | BC061765.1 | - | 1, 2 | 199 | 5620893 | |