siRNA Details
| Gene Symbol: Cadps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013219.1 | NP_037351.1 | 2854 | |||
| GenBank mRNA | U16802.1 | - | 2854 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cadps| Gene Symbol: Cadps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013219.1 | NP_037351.1 | 2854 | |||
| GenBank mRNA | U16802.1 | - | 2854 | |||