siRNA Details
| Gene Symbol: Hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013221.2 | NP_037353.2 | 7 | 903 | ||
| XM_006239975.2 | XP_006240037.1 | 7 | 966 | |||
| GenBank mRNA | BC091165.1 | - | 6 | 877 | 7097013 | |
| FQ223257.1 | - | 7 | 1010 | |||
| FQ224288.1 | - | 7 | 894 | |||
| U09551.1 | - | 6 | 898 | |||

Hbp1
Genomic Map