siRNA Details
| Gene Symbol: Txndc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172032.2 | NP_742029.3 | 2, 3 | 300 | ||
| GenBank mRNA | AF508022.1 | - | 431 | |||
| BC060541.1 | - | 2, 3 | 290 | |||
| BC088106.1 | - | 2, 3 | 300 | 7380129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txndc9| Gene Symbol: Txndc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172032.2 | NP_742029.3 | 2, 3 | 300 | ||
| GenBank mRNA | AF508022.1 | - | 431 | |||
| BC060541.1 | - | 2, 3 | 290 | |||
| BC088106.1 | - | 2, 3 | 300 | 7380129 | ||