siRNA Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100489.1 | NP_001093959.1 | 3 | 305 | ||
| XM_006237179.1 | XP_006237241.1 | 2 | 183 | |||
| GenBank mRNA | BC161860.1 | - | 448 | 7096336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt5b| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100489.1 | NP_001093959.1 | 3 | 305 | ||
| XM_006237179.1 | XP_006237241.1 | 2 | 183 | |||
| GenBank mRNA | BC161860.1 | - | 448 | 7096336 | ||