siRNA Details
Gene Symbol: Mbnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191566.1 | NP_001178495.1 | 7, 8 | 1114 | ||
XM_006232412.2 | XP_006232474.1 | 10, 11 | 2283 | |||
XM_006232430.2 | XP_006232492.1 | 9, 10 | 1625 | |||
XM_006232434.2 | XP_006232496.1 | 7, 8 | 1092 | |||
XM_008761032.1 | XP_008759254.1 | 8, 9 | 1241 | |||
XM_008761033.1 | XP_008759255.1 | 8, 9 | 1223 | |||
XM_008761034.1 | XP_008759256.1 | 7, 8 | 1187 | |||
XM_008761035.1 | XP_008759257.1 | 8, 9 | 1166 | |||
XM_008761037.1 | XP_008759259.1 | 7, 8 | 1146 | |||
XM_008761038.1 | XP_008759260.1 | 7, 8 | 1112 | |||
XM_008761040.1 | XP_008759262.1 | 6, 7 | 1076 | |||
XM_008761042.1 | XP_008759264.1 | 6, 7 | 1035 | |||
XM_008761044.1 | XP_008759266.1 | 5, 6 | 981 |
