siRNA Details
| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172224.1 | NP_757378.1 | 3, 4 | 514 | ||
| XM_006254856.2 | XP_006254918.1 | 3, 4 | 3833 | |||
| GenBank mRNA | AY160191.1 | - | 3, 4 | 514 | ||
| BC083544.1 | - | 3, 4 | 488 | 7191219 | ||

Impa2
Genomic Map