siRNA Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172320.1 | NP_758823.1 | 3, 4 | 262 | ||
| XM_008770001.1 | XP_008768223.1 | 3, 4 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Afm| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172320.1 | NP_758823.1 | 3, 4 | 262 | ||
| XM_008770001.1 | XP_008768223.1 | 3, 4 | 310 | |||