siRNA Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172320.1 | NP_758823.1 | 12, 13 | 1636 | ||
| XM_008770001.1 | XP_008768223.1 | 12, 13 | 1684 | |||
| GenBank mRNA | BC091391.1 | - | 9, 10 | 1201 | ||
| FQ213773.1 | - | 8, 9 | 1072 | |||
| FQ219466.1 | - | 9, 10 | 1170 | |||

Afm
Genomic Map