siRNA Details
Gene Symbol: Slc6a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172321.1 | NP_758824.1 | 11 | 2583 | ||
GenBank mRNA | L22022.1 | - | 11 | 2583 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc6a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172321.1 | NP_758824.1 | 11 | 2583 | ||
GenBank mRNA | L22022.1 | - | 11 | 2583 |