siRNA Details
| Gene Symbol: Trpc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191691.2 | NP_001178620.2 | 8 | 2049 | ||
| GenBank mRNA | AY157999.1 | - | 2 | 164 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpc7| Gene Symbol: Trpc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191691.2 | NP_001178620.2 | 8 | 2049 | ||
| GenBank mRNA | AY157999.1 | - | 2 | 164 | ||