siRNA Details
| Gene Symbol: Elac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172326.1 | NP_758829.1 | 17 | 1588 | ||
| GenBank mRNA | AY149902.1 | - | 17 | 1588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elac2| Gene Symbol: Elac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172326.1 | NP_758829.1 | 17 | 1588 | ||
| GenBank mRNA | AY149902.1 | - | 17 | 1588 | ||