siRNA Details
| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172328.2 | NP_758831.1 | 1 | 105 | ||
| GenBank mRNA | AF521561.1 | - | 1 | 105 | ||
| AY471575.1 | - | 1 | 105 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tac4| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172328.2 | NP_758831.1 | 1 | 105 | ||
| GenBank mRNA | AF521561.1 | - | 1 | 105 | ||
| AY471575.1 | - | 1 | 105 | |||