siRNA Details
| Gene Symbol: Ccr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172329.1 | NP_758832.1 | 3 | 721 | ||
| XM_006244179.2 | XP_006244241.1 | 3 | 1527 | |||
| GenBank mRNA | AF458780.1 | - | 3 | 721 | ||
| BC091115.1 | - | 3 | 735 | 7381553 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ccr9
Genomic Map