siRNA Details
| Gene Symbol: Cthrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271300.1 | NP_001258229.1 | 2 | 406 | ||
| NM_172333.2 | NP_758836.1 | 2 | 451 | |||
| GenBank mRNA | AY136824.1 | - | 2 | 453 | ||
| FQ220442.1 | - | 2 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cthrc1| Gene Symbol: Cthrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271300.1 | NP_001258229.1 | 2 | 406 | ||
| NM_172333.2 | NP_758836.1 | 2 | 451 | |||
| GenBank mRNA | AY136824.1 | - | 2 | 453 | ||
| FQ220442.1 | - | 2 | 363 | |||