siRNA Details
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176857.3 | NP_789827.3 | 1 | 2475 | ||
GenBank mRNA | AB045378.2 | - | 1 | 2475 | ||
AF289091.1 | - | 2533 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176857.3 | NP_789827.3 | 1 | 2475 | ||
GenBank mRNA | AB045378.2 | - | 1 | 2475 | ||
AF289091.1 | - | 2533 |