siRNA Details
Gene Symbol: Cyp4f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173123.1 | NP_775146.1 | 9 | 1239 | ||
GenBank mRNA | U39206.1 | - | 9 | 1239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp4f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173123.1 | NP_775146.1 | 9 | 1239 | ||
GenBank mRNA | U39206.1 | - | 9 | 1239 |