siRNA Details
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004442.1 | NP_001004442.1 | 7 | 679 | ||
GenBank mRNA | BC082063.1 | - | 7 | 679 | 7132589 | |
X79328.1 | - | 6 | 619 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004442.1 | NP_001004442.1 | 7 | 679 | ||
GenBank mRNA | BC082063.1 | - | 7 | 679 | 7132589 | |
X79328.1 | - | 6 | 619 |