siRNA Details
| Gene Symbol: Krt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173128.1 | NP_775151.1 | 1 | 192 | ||
| GenBank mRNA | M63665.1 | - | 1 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt20| Gene Symbol: Krt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173128.1 | NP_775151.1 | 1 | 192 | ||
| GenBank mRNA | M63665.1 | - | 1 | 192 | ||