siRNA Details
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173128.1 | NP_775151.1 | 2 | 456 | ||
GenBank mRNA | M63665.1 | - | 2 | 456 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173128.1 | NP_775151.1 | 2 | 456 | ||
GenBank mRNA | M63665.1 | - | 2 | 456 |