siRNA Details
| Gene Symbol: Phax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173133.2 | NP_775156.2 | 2 | 489 | ||
| GenBank mRNA | BC086410.1 | - | 2 | 459 | ||
| BC097932.1 | - | 2 | 465 | 7386998 | ||
| FQ226770.1 | - | 1 | 37 | |||
| X67877.1 | - | 2 | 489 | |||

Phax
Genomic Map