siRNA Details
| Gene Symbol: Gnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173139.1 | NP_775162.1 | 7 | 905 | ||
| GenBank mRNA | X65747.1 | - | 7 | 905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnat3| Gene Symbol: Gnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173139.1 | NP_775162.1 | 7 | 905 | ||
| GenBank mRNA | X65747.1 | - | 7 | 905 | ||