siRNA Details
Gene Symbol: Gnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173139.1 | NP_775162.1 | 6, 7 | 840 | ||
GenBank mRNA | X65747.1 | - | 6, 7 | 840 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173139.1 | NP_775162.1 | 6, 7 | 840 | ||
GenBank mRNA | X65747.1 | - | 6, 7 | 840 |