siRNA Details
| Gene Symbol: Abi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173143.1 | NP_775166.1 | 8 | 1481 | ||
| GenBank mRNA | FQ222350.1 | - | 1840 | |||
| U94904.1 | - | 8 | 1481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abi2| Gene Symbol: Abi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173143.1 | NP_775166.1 | 8 | 1481 | ||
| GenBank mRNA | FQ222350.1 | - | 1840 | |||
| U94904.1 | - | 8 | 1481 | |||