siRNA Details
| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173146.2 | NP_775169.4 | 399 | |||
| GenBank mRNA | U75361.1 | - | 413 | |||
| Gene Symbol: LOC103693145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766412.1 | XP_008764634.1 | 2 | 1282 | ||
| XM_008766413.1 | XP_008764635.1 | 2 | 1344 | |||
| XM_008766414.1 | XP_008764636.1 | 2 | 755 | |||
| XM_008766415.1 | XP_008764637.1 | 2 | 1162 | |||

Unc13c
Genomic Map