siRNA Details
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173154.1 | NP_775177.1 | 4 | 539 | ||
GenBank mRNA | AF302047.1 | - | 4 | 539 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173154.1 | NP_775177.1 | 4 | 539 | ||
GenBank mRNA | AF302047.1 | - | 4 | 539 |