siRNA Details
| Gene Symbol: Tox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173324.2 | NP_775446.2 | 7 | 1313 | ||
| GenBank mRNA | BC081696.1 | - | 7 | 1313 | 7112549 | |
| FQ210274.1 | - | 7 | 1321 | |||
| FQ212245.1 | - | 7 | 1321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tox4| Gene Symbol: Tox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173324.2 | NP_775446.2 | 7 | 1313 | ||
| GenBank mRNA | BC081696.1 | - | 7 | 1313 | 7112549 | |
| FQ210274.1 | - | 7 | 1321 | |||
| FQ212245.1 | - | 7 | 1321 | |||