siRNA Details
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173324.2 | NP_775446.2 | 9 | 1880 | ||
GenBank mRNA | AF267197.1 | - | 8 | 1832 | ||
BC081696.1 | - | 9 | 1880 | 7112549 | ||
FQ210274.1 | - | 9 | 1888 | |||
FQ212245.1 | - | 9 | 1888 |
