siRNA Details
| Gene Symbol: LOC286992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173326.1 | NP_775448.1 | 432 | |||
| GenBank mRNA | AF012282.1 | - | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC286992| Gene Symbol: LOC286992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173326.1 | NP_775448.1 | 432 | |||
| GenBank mRNA | AF012282.1 | - | 432 | |||