siRNA Details
| Gene Symbol: Slco6c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173338.1 | NP_775460.1 | 708 | |||
| XM_008767361.1 | XP_008765583.1 | 590 | ||||
| GenBank mRNA | AF326113.1 | - | 708 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco6c1| Gene Symbol: Slco6c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173338.1 | NP_775460.1 | 708 | |||
| XM_008767361.1 | XP_008765583.1 | 590 | ||||
| GenBank mRNA | AF326113.1 | - | 708 | |||