siRNA Details
| Gene Symbol: Tnfrsf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105761.1 | NP_001099231.1 | 3 | 579 | ||
| XM_008767480.1 | XP_008765702.1 | 3 | 588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf17| Gene Symbol: Tnfrsf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105761.1 | NP_001099231.1 | 3 | 579 | ||
| XM_008767480.1 | XP_008765702.1 | 3 | 588 | |||