siRNA Details
| Gene Symbol: Trap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039001.1 | NP_001034090.1 | 2 | 182 | ||
| GenBank mRNA | BC083909.1 | - | 2 | 182 | 7134204 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trap1| Gene Symbol: Trap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039001.1 | NP_001034090.1 | 2 | 182 | ||
| GenBank mRNA | BC083909.1 | - | 2 | 182 | 7134204 | |