siRNA Details
| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105763.1 | NP_001099233.1 | 4 | 993 | ||
| XM_006245830.2 | XP_006245892.1 | 4 | 5665 | |||
| XM_006245831.2 | XP_006245893.1 | 2 | 5409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp263| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105763.1 | NP_001099233.1 | 4 | 993 | ||
| XM_006245830.2 | XP_006245892.1 | 4 | 5665 | |||
| XM_006245831.2 | XP_006245893.1 | 2 | 5409 | |||