siRNA Details
| Gene Symbol: Tbc1d24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105769.1 | NP_001099239.1 | 4, 5 | 1396 | ||
| XM_006245926.1 | XP_006245988.1 | 3, 4 | 1155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d24| Gene Symbol: Tbc1d24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105769.1 | NP_001099239.1 | 4, 5 | 1396 | ||
| XM_006245926.1 | XP_006245988.1 | 3, 4 | 1155 | |||