siRNA Details
Gene Symbol: Tbc1d24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105769.1 | NP_001099239.1 | 3 | 890 | ||
XM_006245926.1 | XP_006245988.1 | 1 | 631 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbc1d24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105769.1 | NP_001099239.1 | 3 | 890 | ||
XM_006245926.1 | XP_006245988.1 | 1 | 631 |