siRNA Details
| Gene Symbol: Zfp598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105770.1 | NP_001099240.1 | 11 | 2286 | ||
| XM_006245934.2 | XP_006245996.1 | 11 | 2334 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp598| Gene Symbol: Zfp598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105770.1 | NP_001099240.1 | 11 | 2286 | ||
| XM_006245934.2 | XP_006245996.1 | 11 | 2334 | |||