siRNA Details
| Gene Symbol: Tbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008277.1 | NP_001008278.1 | 15, 16 | 1735 | ||
| GenBank mRNA | BC085837.1 | - | 15, 16 | 1735 | 7135552 | |
More Information
Other Species Targeted
Set Membership: |
|

Tbl3
Genomic Map